Product Details
- SNP ID
-
hCV8729307
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:16587382 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGATACAACTTTTTCCCTTAGAA[A/G]GGTATTATATCCCATTGCTACCGTT
- Phenotype
-
MIM: 601486
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DAZL
PubMed Links
Gene Details
- Gene
- DAZL
- Gene Name
- deleted in azoospermia like
View Full Product Details