Product Details

SNP ID
rs1754437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:94604439 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGACCCAAATACATCCAAACACA[C/T]AGGTTTTGTGTTTAAGGGAACTGTT
Phenotype
MIM: 611570
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBP1 PubMed Links

Gene Details

Gene
FBP1
Gene Name
fructose-bisphosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000507.3 Intron NP_000498.2
NM_001127628.1 Intron NP_001121100.1
XM_006717005.3 Intron XP_006717068.1

View Full Product Details