Product Details

SNP ID
rs1061858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:78658113 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGTGGATGTTGTTGGCGCTGTCT[G/T]ACAGTTCTGGGTACTGCTCGACAGA
Phenotype
MIM: 610084
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TENM4 PubMed Links

Gene Details

Gene
TENM4
Gene Name
teneurin transmembrane protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098816.2 5813 Nonsense Mutation TAA,TCA *2752S NP_001092286.2
XM_011544933.2 5813 Nonsense Mutation TAA,TCA *1900S XP_011543235.1
XM_017017525.1 5813 Nonsense Mutation TAA,TCA *2777S XP_016873014.1
XM_017017526.1 5813 Nonsense Mutation TAA,TCA *2752S XP_016873015.1

View Full Product Details