Product Details

SNP ID
rs1049886
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143612680 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGCAGGATCAGAGTGGACACGC[C/G]CAGGGGGCTGGTGTGGGGAGCAAAG
Phenotype
MIM: 616408 MIM: 137020
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
PYCRL PubMed Links

Gene Details

Gene
PYCRL
Gene Name
pyrroline-5-carboxylate reductase-like
There are no transcripts associated with this gene.

Gene
TSTA3
Gene Name
tissue specific transplantation antigen P35B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317783.1 1323 UTR 3 NP_001304712.1
NM_003313.3 1323 UTR 3 NP_003304.1
XM_005251051.3 1323 UTR 3 XP_005251108.2
XM_011517269.1 1323 UTR 3 XP_011515571.1

View Full Product Details