Product Details

SNP ID
rs1333476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:44946754 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGAAGAAACAGATTGTTCCTATA[G/T]ATTTAGAATCCAGGTAAATAAGCAG
Phenotype
MIM: 604354
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NUFIP1 PubMed Links

Gene Details

Gene
NUFIP1
Gene Name
NUFIP1, FMR1 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012345.2 Intron NP_036477.2

View Full Product Details