Product Details

SNP ID
rs3448
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:49359318 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAGAGCACTGTGAATTAGAGCCAGA[C/T]GCTTAAGTCCAGGTGAGACAGGTTA
Phenotype
MIM: 138320 MIM: 165390
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPX1 PubMed Links

Gene Details

Gene
GPX1
Gene Name
glutathione peroxidase 1
There are no transcripts associated with this gene.

Gene
RHOA
Gene Name
ras homolog family member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001313941.1 1630 UTR 3 NP_001300870.1
NM_001313943.1 1630 UTR 3 NP_001300872.1
NM_001313944.1 1630 UTR 3 NP_001300873.1
NM_001313945.1 1630 UTR 3 NP_001300874.1
NM_001313946.1 1630 UTR 3 NP_001300875.1
NM_001313947.1 1630 UTR 3 NP_001300876.1
NM_001664.3 1630 UTR 3 NP_001655.1

View Full Product Details