Product Details

SNP ID
rs2343144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:32396642 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTATGTAAGTAGACCTGTGGATG[A/T]GTGTGAGGGGGCAAGGTGCTCTGCA
Phenotype
MIM: 607890
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CMTM7 PubMed Links

Gene Details

Gene
CMTM7
Gene Name
CKLF like MARVEL transmembrane domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138410.3 Intron NP_612419.1
NM_181472.2 Intron NP_852137.1
XM_011533319.2 Intron XP_011531621.1
XM_017005646.1 Intron XP_016861135.1

View Full Product Details