Product Details

SNP ID
rs1064247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:180790887 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAACAGGGAGGGCACTGATTTTCA[A/G]AAAGGAGGGGTCAGGGCATAGTCCC
Phenotype
MIM: 160995
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MGAT1 PubMed Links

Gene Details

Gene
MGAT1
Gene Name
mannosyl (alpha-1,3-)-glycoprotein beta-1,2-N-acetylglucosaminyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114617.1 2851 UTR 3 NP_001108089.1
NM_001114618.1 2851 UTR 3 NP_001108090.1
NM_001114619.1 2851 UTR 3 NP_001108091.1
NM_001114620.1 2851 UTR 3 NP_001108092.1
NM_002406.3 2851 UTR 3 NP_002397.2
XM_005265915.1 2851 UTR 3 XP_005265972.1
XM_005265916.1 2851 UTR 3 XP_005265973.1
XM_006714866.1 2851 UTR 3 XP_006714929.1
XM_011534559.1 2851 UTR 3 XP_011532861.1
XM_011534560.1 2851 UTR 3 XP_011532862.1
XM_011534561.1 2851 UTR 3 XP_011532863.1
XM_011534562.1 2851 UTR 3 XP_011532864.1
XM_011534563.2 2851 UTR 3 XP_011532865.1
XM_017009486.1 2851 UTR 3 XP_016864975.1
XM_017009487.1 2851 UTR 3 XP_016864976.1

View Full Product Details