Product Details

SNP ID
rs1043071
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:72354578 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTACACATCTGCAGGTCTAACTCA[A/G]AACTAAGGTACATAAAATTGAAAGC
Phenotype
MIM: 604761
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZFAND5 PubMed Links

Gene Details

Gene
ZFAND5
Gene Name
zinc finger AN1-type containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102420.2 2294 UTR 3 NP_001095890.1
NM_001102421.2 2294 UTR 3 NP_001095891.1
NM_001278243.1 2294 UTR 3 NP_001265172.1
NM_001278244.1 2294 UTR 3 NP_001265173.1
NM_001278245.1 2294 UTR 3 NP_001265174.1
NM_006007.3 2294 UTR 3 NP_005998.1

View Full Product Details