Product Details

SNP ID
rs1042375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:113388236 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAAAAGCAGCATTTACTTTGGTT[G/T]TCACTTGTCTGAGGCCCCGGGAACG
Phenotype
MIM: 125270
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ALAD PubMed Links

Gene Details

Gene
ALAD
Gene Name
aminolevulinate dehydratase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000031.5 1425 UTR 3 NP_000022.3
NM_001003945.2 1425 UTR 3 NP_001003945.1
NM_001317745.1 1425 UTR 3 NP_001304674.1
XM_011518364.2 1425 UTR 3 XP_011516666.1

View Full Product Details