Product Details

SNP ID
rs1047426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:10441956 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAACTGCAGGGTCTAGGGTGTTGT[C/T]GGAGTGGCAGTTGGTCCGAATTTCT
Phenotype
MIM: 613297 MIM: 611756
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MARCH6 PubMed Links

Gene Details

Gene
MARCH6
Gene Name
membrane associated ring-CH-type finger 6
There are no transcripts associated with this gene.

Gene
ROPN1L
Gene Name
rhophilin associated tail protein 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201466.1 94 UTR 5 NP_001188395.1
NM_031916.4 94 UTR 5 NP_114122.2
XM_006714503.2 94 UTR 5 XP_006714566.1
XM_006714504.2 94 UTR 5 XP_006714567.1
XM_017009946.1 94 UTR 5 XP_016865435.1
XM_017009947.1 94 UTR 5 XP_016865436.1
Gene
ROPN1L-AS1
Gene Name
ROPN1L antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details