Product Details

SNP ID
rs1053116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:40760729 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGCATGATTCTAGAAGTGCTTGA[C/G]TCAAATCTACTAGAGTCACCATTTA
Phenotype
MIM: 602739
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRKAA1 PubMed Links

Gene Details

Gene
PRKAA1
Gene Name
protein kinase AMP-activated catalytic subunit alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006251.5 4691 UTR 3 NP_006242.5
NM_206907.3 4691 UTR 3 NP_996790.3
XM_006714482.1 4691 UTR 3 XP_006714545.1
XM_017009624.1 4691 UTR 3 XP_016865113.1
XM_017009625.1 4691 UTR 3 XP_016865114.1
Gene
TTC33
Gene Name
tetratricopeptide repeat domain 33
There are no transcripts associated with this gene.

View Full Product Details