Product Details
- SNP ID
-
rs1012356
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:68250838 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACCAATTCAGACTTCCATTTAACT[A/T]TAATAAATCTCTTACTAATATTTTT
- Phenotype
-
MIM: 605330
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IL22
PubMed Links
Gene Details
- Gene
- IL22
- Gene Name
- interleukin 22
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020525.4 |
|
Intron |
|
|
NP_065386.1 |
View Full Product Details