Product Details

SNP ID
rs1376320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:42897897 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAATTAATTGTGCCCAAAAGTTT[A/G]TAAAGTGAACAGAAAAGTTATTACT
Phenotype
MIM: 613283
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRXCR1 PubMed Links

Gene Details

Gene
GRXCR1
Gene Name
glutaredoxin and cysteine rich domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080476.2 Intron NP_001073945.1

View Full Product Details