Product Details

SNP ID
rs904326
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:219912946 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATACAGGAGCACCAAACTGAGTG[C/T]CAGTGTTTTTCTAATCTTAAAGGAA
Phenotype
MIM: 611146
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC30A10 PubMed Links

Gene Details

Gene
SLC30A10
Gene Name
solute carrier family 30 member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018713.2 4508 Intron NP_061183.2
XM_006711437.3 4508 UTR 3 XP_006711500.2
XM_017001684.1 4508 UTR 3 XP_016857173.1

View Full Product Details