Product Details

SNP ID
rs131159
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:44496060 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CTGCCCAATGGCTGAGAGTCCACCA[C/T]GACCTTATTTAGCAGGCAAGTTCCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LDOC1L PubMed Links

Gene Details

Gene
LDOC1L
Gene Name
leucine zipper down-regulated in cancer 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032287.2 1834 UTR 3 NP_115663.2

View Full Product Details