Product Details

SNP ID
rs798180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:15838464 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGTAGGTGAAAAAAGGACTCAAA[C/T]GTGACATTTTTCAGAAAGTATTTTT
Phenotype
MIM: 300629
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AP1S2 PubMed Links

Gene Details

Gene
AP1S2
Gene Name
adaptor related protein complex 1 sigma 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272071.1 Intron NP_001259000.1
NM_003916.4 Intron NP_003907.3
XM_005274614.3 Intron XP_005274671.1
XM_011545599.1 Intron XP_011543901.1
XM_017029925.1 Intron XP_016885414.1
XM_017029926.1 Intron XP_016885415.1

View Full Product Details