Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001127391.2 | 1346 | UTR 3 | NP_001120863.1 | ||
NM_001289993.1 | 1346 | UTR 3 | NP_001276922.1 | ||
NM_139163.3 | 1346 | UTR 3 | NP_631902.2 | ||
XM_011510606.2 | 1346 | UTR 3 | XP_011508908.1 | ||
XM_011510610.2 | 1346 | UTR 3 | XP_011508912.1 | ||
XM_011510612.2 | 1346 | UTR 3 | XP_011508914.1 | ||
XM_017003360.1 | 1346 | Intron | XP_016858849.1 | ||
XM_017003361.1 | 1346 | Intron | XP_016858850.1 | ||
XM_017003362.1 | 1346 | UTR 3 | XP_016858851.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001080124.1 | 1346 | Intron | NP_001073593.1 | ||
NM_001080125.1 | 1346 | Intron | NP_001073594.1 | ||
NM_001228.4 | 1346 | Intron | NP_001219.2 | ||
NM_033355.3 | 1346 | Intron | NP_203519.1 | ||
NM_033356.3 | 1346 | Intron | NP_203520.1 | ||
NM_033358.3 | 1346 | Intron | NP_203522.1 | ||
XM_005246885.1 | 1346 | Intron | XP_005246942.1 | ||
XM_005246886.1 | 1346 | Intron | XP_005246943.1 | ||
XM_005246887.1 | 1346 | Intron | XP_005246944.1 | ||
XM_005246888.1 | 1346 | Intron | XP_005246945.1 | ||
XM_005246889.1 | 1346 | Intron | XP_005246946.1 | ||
XM_005246890.3 | 1346 | Intron | XP_005246947.1 | ||
XM_005246891.4 | 1346 | Intron | XP_005246948.1 | ||
XM_005246892.1 | 1346 | Intron | XP_005246949.1 | ||
XM_005246893.2 | 1346 | Intron | XP_005246950.1 | ||
XM_005246894.3 | 1346 | Intron | XP_005246951.1 | ||
XM_005246895.2 | 1346 | Intron | XP_005246952.1 | ||
XM_006712789.1 | 1346 | Intron | XP_006712852.1 | ||
XM_006712790.3 | 1346 | Intron | XP_006712853.1 | ||
XM_006712793.2 | 1346 | Intron | XP_006712856.1 | ||
XM_011511969.1 | 1346 | Intron | XP_011510271.1 |