Product Details

SNP ID
rs14272
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:58113783 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCTCCCCCCACACTGGGGCAGGC[A/G]GCGGAATAAGCTCCAGCGTTCATGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFAP20 PubMed Links

Gene Details

Gene
CFAP20
Gene Name
cilia and flagella associated protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013242.2 1101 UTR 3 NP_037374.1

View Full Product Details