Product Details

SNP ID
rs1045433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:3149742 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAATTTTCATTTACTCTATTTACA[C/T]AAATATGCAGTGTGACACTCAACTG
Phenotype
MIM: 609262 MIM: 612907
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRBN PubMed Links

Gene Details

Gene
CRBN
Gene Name
cereblon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173482.1 2479 UTR 3 NP_001166953.1
NM_016302.3 2479 UTR 3 NP_057386.2
XM_005265202.3 2479 UTR 3 XP_005265259.1
XM_011533791.2 2479 UTR 3 XP_011532093.1
XM_011533793.2 2479 UTR 3 XP_011532095.1
XM_011533794.2 2479 UTR 3 XP_011532096.1
Gene
TRNT1
Gene Name
tRNA nucleotidyl transferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302946.1 2479 Intron NP_001289875.1
NM_182916.2 2479 Intron NP_886552.2
XM_011533776.2 2479 Intron XP_011532078.1
XM_011533777.2 2479 Intron XP_011532079.1
XM_011533778.2 2479 Intron XP_011532080.1

View Full Product Details