Product Details

SNP ID
rs1802412
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:28111163 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTTCACAGAAAGGCCTCAATTC[A/T]GATGACATAATGCATATGCTACAAG
Phenotype
MIM: 605837
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HERC2 PubMed Links

Gene Details

Gene
HERC2
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004667.5 8543 UTR 3 NP_004658.3
XM_005268276.4 8543 UTR 3 XP_005268333.1
XM_006720726.3 8543 UTR 3 XP_006720789.1
XM_006720727.3 8543 UTR 3 XP_006720790.1
XM_011522131.2 8543 UTR 3 XP_011520433.1
XM_011522133.2 8543 UTR 3 XP_011520435.1
XM_017022695.1 8543 UTR 3 XP_016878184.1
XM_017022696.1 8543 UTR 3 XP_016878185.1
XM_017022697.1 8543 UTR 3 XP_016878186.1
XM_017022698.1 8543 UTR 3 XP_016878187.1

View Full Product Details