Product Details

SNP ID
rs13287
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102480681 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGGCCTCAGGCCATCACCACTGC[A/G]GGTAGTTCATCGTCCTTTCTGGGCC
Phenotype
MIM: 605143
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTR1A PubMed Links

Gene Details

Gene
ACTR1A
Gene Name
ARP1 actin-related protein 1 homolog A, centractin alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005736.3 1421 UTR 3 NP_005727.1
Gene
MFSD13A
Gene Name
major facilitator superfamily domain containing 13A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024789.3 1421 Intron NP_079065.2
XM_006717973.3 1421 Intron XP_006718036.1
XM_006717974.3 1421 Intron XP_006718037.1
XM_011540160.2 1421 Intron XP_011538462.1
XM_011540163.2 1421 Intron XP_011538465.1
XM_011540167.2 1421 Intron XP_011538469.1
XM_011540169.2 1421 Intron XP_011538471.1
XM_017016654.1 1421 Intron XP_016872143.1
XM_017016655.1 1421 Intron XP_016872144.1
XM_017016656.1 1421 Intron XP_016872145.1
XM_017016657.1 1421 Intron XP_016872146.1
XM_017016658.1 1421 Intron XP_016872147.1
XM_017016659.1 1421 Intron XP_016872148.1
XM_017016660.1 1421 Intron XP_016872149.1
XM_017016661.1 1421 Intron XP_016872150.1
XM_017016662.1 1421 Intron XP_016872151.1

View Full Product Details