Product Details

SNP ID
rs1029169
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:110500546 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCTGGTGGCATCTCAGATCTTGG[C/T]GGGGGCAATGCCTTTTGACACTTTC
Phenotype
MIM: 603069
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP5 PubMed Links

Gene Details

Gene
DUSP5
Gene Name
dual specificity phosphatase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004419.3 Intron NP_004410.3

View Full Product Details