Product Details

SNP ID
rs1112962
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:27800413 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCATTTGCTGAGTTTCCACTGAATC[A/G]GTTCAGTAACCAGAAGAAAGTTCAA
Phenotype
MIM: 615408
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARMC4 PubMed Links

Gene Details

Gene
ARMC4
Gene Name
armadillo repeat containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290020.1 Intron NP_001276949.1
NM_001290021.1 Intron NP_001276950.1
NM_001312689.1 Intron NP_001299618.1
NM_018076.4 Intron NP_060546.2
XM_011519527.1 Intron XP_011517829.1
XM_011519528.1 Intron XP_011517830.1
XM_011519533.2 Intron XP_011517835.1
XM_011519534.2 Intron XP_011517836.1
XM_011519535.2 Intron XP_011517837.1
XM_011519537.2 Intron XP_011517839.1
XM_017016368.1 Intron XP_016871857.1
XM_017016369.1 Intron XP_016871858.1
XM_017016370.1 Intron XP_016871859.1
XM_017016371.1 Intron XP_016871860.1

View Full Product Details