Product Details
- SNP ID
-
rs3914138
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:71128836 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGAGACAATTACCTCTACTTTTTA[A/G]AACTTCTTGGATTTCTCTAGAATGT
- Phenotype
-
MIM: 600769
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TSPAN8
PubMed Links
Gene Details
- Gene
- TSPAN8
- Gene Name
- tetraspanin 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004616.2 |
|
Intron |
|
|
NP_004607.1 |
View Full Product Details