Product Details

SNP ID
rs618703
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:71120493 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGGGCGTGGTCCGAGGGCAGTGA[A/G]CCAGGTTCACAGTGCCGCTGTGTCT
Phenotype
MIM: 614476
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CT62 PubMed Links

Gene Details

Gene
CT62
Gene Name
cancer/testis antigen 62
There are no transcripts associated with this gene.

Gene
THSD4
Gene Name
thrombospondin type 1 domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286429.1 Intron NP_001273358.1
NM_024817.2 Intron NP_079093.2
XM_006720692.3 Intron XP_006720755.1
XM_011522043.2 Intron XP_011520345.1
XM_011522044.2 Intron XP_011520346.1
XM_017022582.1 Intron XP_016878071.1
XM_017022583.1 Intron XP_016878072.1
XM_017022584.1 Intron XP_016878073.1
XM_017022585.1 Intron XP_016878074.1
XM_017022586.1 Intron XP_016878075.1

View Full Product Details