Product Details

SNP ID
rs1046159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74179559 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAAAAATATCTGGCTCTCGAGTG[G/T]GGGCAGCTCAGTGTGGGACCTGGTC
Phenotype
MIM: 602059 MIM: 610745
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ISLR PubMed Links

Gene Details

Gene
ISLR
Gene Name
immunoglobulin superfamily containing leucine rich repeat
There are no transcripts associated with this gene.

Gene
STRA6
Gene Name
stimulated by retinoic acid 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142617.1 2737 UTR 3 NP_001136089.1
NM_001142618.1 2737 UTR 3 NP_001136090.1
NM_001142619.1 2737 UTR 3 NP_001136091.1
NM_001142620.1 2737 Intron NP_001136092.1
NM_001199040.1 2737 UTR 3 NP_001185969.1
NM_001199041.1 2737 UTR 3 NP_001185970.1
NM_001199042.1 2737 UTR 3 NP_001185971.1
NM_022369.3 2737 UTR 3 NP_071764.3
XM_011521883.1 2737 UTR 3 XP_011520185.1
XM_011521884.1 2737 UTR 3 XP_011520186.1
XM_011521885.2 2737 Intron XP_011520187.1
XM_017022478.1 2737 UTR 3 XP_016877967.1
XM_017022479.1 2737 UTR 3 XP_016877968.1
XM_017022480.1 2737 UTR 3 XP_016877969.1

View Full Product Details