Product Details

SNP ID
rs1396794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:69645350 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCCAGGGTGAAATGCAGCCCCT[A/C]TTCCCTCAAACTAAGCCCACGAAGG
Phenotype
MIM: 616405 MIM: 106491
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AAK1 PubMed Links

Gene Details

Gene
AAK1
Gene Name
AP2 associated kinase 1
There are no transcripts associated with this gene.

Gene
ANXA4
Gene Name
annexin A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001153.4 Intron NP_001144.1
NM_001320698.1 Intron NP_001307627.1
NM_001320700.1 Intron NP_001307629.1
NM_001320702.1 Intron NP_001307631.1
XM_017003943.1 Intron XP_016859432.1
XM_017003944.1 Intron XP_016859433.1

View Full Product Details