Product Details

SNP ID
rs1447514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:22340750 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGCTGTGGAGCCCTTTGTCTCCAA[C/T]TCGAGTAATAAGAAAAAAAAAATAG
Phenotype
MIM: 607563
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC17A6 PubMed Links

Gene Details

Gene
SLC17A6
Gene Name
solute carrier family 17 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020346.2 Intron NP_065079.1

View Full Product Details