Product Details

SNP ID
rs2864
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:138187741 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAGAAAAATTAAGAATCAAACATCA[C/T]TCTGGACCATGGGAACCTTGAAAAG
Phenotype
MIM: 602848 MIM: 603462 MIM: 605664
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BRD8 PubMed Links

Gene Details

Gene
BRD8
Gene Name
bromodomain containing 8
There are no transcripts associated with this gene.

Gene
CDC23
Gene Name
cell division cycle 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004661.3 3062 UTR 3 NP_004652.2
Gene
KIF20A
Gene Name
kinesin family member 20A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005733.2 3062 Intron NP_005724.1

View Full Product Details