Product Details

SNP ID
rs832193
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:63868769 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCACTTATAATCTGAATTAAAAAA[C/T]TAAAATACTGCACATGTACTTTATA
Phenotype
MIM: 607640 MIM: 611965
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATXN7 PubMed Links

Gene Details

Gene
ATXN7
Gene Name
ataxin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000333.3 Intron NP_000324.1
NM_001128149.2 Intron NP_001121621.2
NM_001177387.1 Intron NP_001170858.1
Gene
THOC7
Gene Name
THO complex 7
There are no transcripts associated with this gene.

Gene
THOC7-AS1
Gene Name
THOC7 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details