Product Details
- SNP ID
-
rs704371
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:63881852 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TTGGGCCAGAGAGTAAAGTAAAGGG[A/G]CTTACCGGCTGCTGGGCCAGGCTAG
- Phenotype
-
MIM: 607640
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ATXN7
PubMed Links
Gene Details
- Gene
- ATXN7
- Gene Name
- ataxin 7
View Full Product Details