Product Details

SNP ID
rs959
Assay Type
Functionally tested
NCBI dbSNP Submissions
60
Location
Chr.1:70852578 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAAAAAAATGCAAATTTTACATA[C/T]GATAATGTATACGCAAATATAAATT
Phenotype
MIM: 176806
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTGER3 PubMed Links

Gene Details

Gene
PTGER3
Gene Name
prostaglandin E receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126044.1 1688 Intron NP_001119516.1
NM_198714.1 1688 UTR 3 NP_942007.1
NM_198715.2 1688 Intron NP_942008.1
NM_198716.1 1688 UTR 3 NP_942009.1
NM_198717.1 1688 UTR 3 NP_942010.1
NM_198718.1 1688 Intron NP_942011.1
NM_198719.1 1688 Intron NP_942012.1
XM_011541810.2 1688 UTR 3 XP_011540112.1

View Full Product Details