Product Details

SNP ID
rs1569824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116269639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCCTAATGATTCTAGACATATGG[A/T]ATTTTGGGCTGATAGGGAAAACAAC
Phenotype
MIM: 605942 MIM: 604714
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DSE PubMed Links

Gene Details

Gene
DSE
Gene Name
dermatan sulfate epimerase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080976.2 Intron NP_001074445.1
NM_001322937.1 Intron NP_001309866.1
NM_001322938.1 Intron NP_001309867.1
NM_001322939.1 Intron NP_001309868.1
NM_001322940.1 Intron NP_001309869.1
NM_001322941.1 Intron NP_001309870.1
NM_001322943.1 Intron NP_001309872.1
NM_001322944.1 Intron NP_001309873.1
NM_013352.3 Intron NP_037484.1
XM_017010795.1 Intron XP_016866284.1
XM_017010796.1 Intron XP_016866285.1
XM_017010797.1 Intron XP_016866286.1
Gene
TSPYL1
Gene Name
TSPY like 1
There are no transcripts associated with this gene.

View Full Product Details