Product Details

SNP ID
rs910391
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116254181 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCGCGCGAGAACTCTCTGCGCGG[G/T]TGACTACGCTTATCAATCCATCGTA
Phenotype
MIM: 605942
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DSE PubMed Links

Gene Details

Gene
DSE
Gene Name
dermatan sulfate epimerase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080976.2 30 Intron NP_001074445.1
NM_001322937.1 30 UTR 5 NP_001309866.1
NM_001322938.1 30 UTR 5 NP_001309867.1
NM_001322939.1 30 Intron NP_001309868.1
NM_001322940.1 30 UTR 5 NP_001309869.1
NM_001322941.1 30 Intron NP_001309870.1
NM_001322943.1 30 Intron NP_001309872.1
NM_001322944.1 30 Intron NP_001309873.1
NM_013352.3 30 Intron NP_037484.1
XM_017010795.1 30 Intron XP_016866284.1
XM_017010796.1 30 Intron XP_016866285.1
XM_017010797.1 30 Intron XP_016866286.1
Gene
NT5DC1
Gene Name
5'-nucleotidase domain containing 1
There are no transcripts associated with this gene.

Gene
TSPYL4
Gene Name
TSPY like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021648.4 30 Intron NP_067680.3

View Full Product Details