Product Details

SNP ID
rs1030869
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:55481709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAATGCAGTGGGGGCTTAAGAAGA[C/T]AACTCTGGACTTAGACCGCTTGGCT
Phenotype
MIM: 120360
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MMP2 PubMed Links

Gene Details

Gene
MMP2
Gene Name
matrix metallopeptidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127891.2 153 UTR 5 NP_001121363.1
NM_001302508.1 153 Intron NP_001289437.1
NM_001302509.1 153 Intron NP_001289438.1
NM_001302510.1 153 Intron NP_001289439.1
NM_004530.5 153 Intron NP_004521.1

View Full Product Details