Product Details

SNP ID
rs10405
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:167541302 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGAAGATGTGATTCTTTTAATGC[A/C]ATGTTCTCTTCCCTCTGGTGACCTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CREG1 PubMed Links

Gene Details

Gene
CREG1
Gene Name
cellular repressor of E1A stimulated genes 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003851.2 1737 UTR 3 NP_003842.1

View Full Product Details