Product Details

SNP ID
rs1558335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6492668 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCATTTATTAACTTTGGAAAAA[C/T]TGTTTATTTTTACTTATGAAGTAAA
Phenotype
MIM: 611855 MIM: 615638
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL51 PubMed Links

Gene Details

Gene
MRPL51
Gene Name
mitochondrial ribosomal protein L51
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016497.3 Intron NP_057581.2
Gene
NCAPD2
Gene Name
non-SMC condensin I complex subunit D2
There are no transcripts associated with this gene.

View Full Product Details