Product Details

SNP ID
rs868478
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:104347196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTCCTGGTAAAAATGCTTACCC[A/G]CTGTGGCTGCTCCCTCAGTGCTTCT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CFAP58 PubMed Links

Gene Details

Gene
CFAP58
Gene Name
cilia and flagella associated protein 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008723.1 Intron NP_001008723.1
XM_011539377.2 Intron XP_011537679.1
XM_017015787.1 Intron XP_016871276.1
XM_017015788.1 Intron XP_016871277.1
XM_017015789.1 Intron XP_016871278.1
XM_017015790.1 Intron XP_016871279.1
Gene
CFAP58-AS1
Gene Name
CFAP58 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
ITPRIP
Gene Name
inositol 1,4,5-trisphosphate receptor interacting protein
There are no transcripts associated with this gene.

View Full Product Details