Product Details

SNP ID
rs7546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4797316 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTGGCACTTCCAGTGTCCATTCC[C/T]GGCAGTGCAAATGTGTTAGGTGGGG
Phenotype
MIM: 610660 MIM: 614574 MIM: 611562
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLYR1 PubMed Links

Gene Details

Gene
GLYR1
Gene Name
glyoxylate reductase 1 homolog
There are no transcripts associated with this gene.

Gene
ROGDI
Gene Name
rogdi homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024589.2 1387 UTR 3 NP_078865.1
XM_006720947.3 1387 UTR 3 XP_006721010.1
XM_006720948.3 1387 UTR 3 XP_006721011.1
Gene
SEPT12
Gene Name
septin 12
There are no transcripts associated with this gene.

Gene
SMIM22
Gene Name
small integral membrane protein 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253790.1 1387 Intron NP_001240719.1
NM_001253791.1 1387 Intron NP_001240720.1
NM_001253793.1 1387 Intron NP_001240722.1
NM_001253794.1 1387 Intron NP_001240723.1
XM_011522499.2 1387 Intron XP_011520801.1
XM_011522500.2 1387 Intron XP_011520802.1
XM_011522501.2 1387 Intron XP_011520803.1

View Full Product Details