Product Details

SNP ID
rs33659
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:11869504 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTAATGGCACAGTTTGATAAGGTA[A/G]GCCAACAATGAATATAACATTTAAC
Phenotype
MIM: 139259
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSPT1 PubMed Links

Gene Details

Gene
GSPT1
Gene Name
G1 to S phase transition 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130006.1 5777 UTR 3 NP_001123478.1
NM_001130007.1 5777 UTR 3 NP_001123479.1
NM_002094.3 5777 UTR 3 NP_002085.2

View Full Product Details