Product Details
- SNP ID
-
rs1048364
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:100749656 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTATAAAATGGTAGGCAATCTCAT[C/T]GTGCATTATCTTTTTGTGCTCAGAC
- Phenotype
-
MIM: 606279
MIM: 602498
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ABI3BP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs573536899] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ABI3BP
- Gene Name
- ABI family member 3 binding protein
- Gene
- TFG
- Gene Name
- TRK-fused gene
View Full Product Details