Product Details

SNP ID
rs3853054
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.Y:8811802 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGAAAGATTATAAGGGAACCTGATT[T/G]CTTGGGAGTGATAATTCAGGAGAAA
Phenotype
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
TTTY11 PubMed Links

Gene Details

Gene
TTTY11
Gene Name
testis-specific transcript, Y-linked 11 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details