Product Details
- SNP ID
-
rs12567
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:108043378 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TGTTATACATCCAAGAGCCTTATAT[C/T]TCACTCCACATAAACAGTAAACATA
- Phenotype
-
MIM: 607649
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OSTM1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73504439,rs9372179] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OSTM1
- Gene Name
- osteopetrosis associated transmembrane protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014028.3 |
2498 |
UTR 3 |
|
|
NP_054747.2 |
View Full Product Details