Product Details

SNP ID
rs1538347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:136827019 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTGGGCTTCAGTTGTACTAACA[C/T]ACATGTATATTTTCAGTGAACATAA
Phenotype
MIM: 601757
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PEX7 PubMed Links

Gene Details

Gene
PEX7
Gene Name
peroxisomal biogenesis factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000288.3 Intron NP_000279.1
XM_005267019.4 Intron XP_005267076.1
XM_006715502.1 Intron XP_006715565.1
XM_017010934.1 Intron XP_016866423.1

View Full Product Details