Product Details

SNP ID
rs1076430
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:121884495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACATCATTATGTTTAATTGTAGC[A/C]TATATTCTGGGTATTTTGAGCCTGA
Phenotype
MIM: 176891
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PTPRZ1 PubMed Links

Gene Details

Gene
PTPRZ1
Gene Name
protein tyrosine phosphatase, receptor type Z1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206838.1 Intron NP_001193767.1
NM_001206839.1 Intron NP_001193768.1
NM_002851.2 Intron NP_002842.2
XM_005250519.1 Intron XP_005250576.1
XM_017012477.1 Intron XP_016867966.1

View Full Product Details