Product Details

SNP ID
rs916440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41971833 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCAGCTCCATCTCCTTGACTCTCC[A/G]TCAGAACAGCCAGGCACGGTCCCTG
Phenotype
MIM: 608314
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEPT3 PubMed Links

Gene Details

Gene
SEPT3
Gene Name
septin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019106.5 583 Intron NP_061979.3
NM_145733.2 583 Intron NP_663786.2
XM_017028862.1 583 Missense Mutation CAT,CGT H114R XP_016884351.1

View Full Product Details