Product Details
- SNP ID
-
rs10421297
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:23916178 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGGCCTCAAGTCTGCCCCTCATCCC[C/T]TATTCCTCCAGCCTGACTGGCTTGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF726
PubMed Links
Gene Details
- Gene
- ZNF726
- Gene Name
- zinc finger protein 726
View Full Product Details