Product Details

SNP ID
rs6982146
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:134598874 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCATCCAAGGATGGGTAAACTGAG[A/G]CAAATAGGTCACATGACTTTTCCCA
Phenotype
MIM: 610931
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZFAT PubMed Links

Gene Details

Gene
ZFAT
Gene Name
zinc finger and AT-hook domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029939.3 Intron NP_001025110.2
NM_001167583.2 Intron NP_001161055.1
NM_001174157.1 Intron NP_001167628.1
NM_001174158.1 Intron NP_001167629.1
NM_001289394.1 Intron NP_001276323.1
NM_020863.3 Intron NP_065914.2
XM_011517203.1 Intron XP_011515505.1
XM_011517204.2 Intron XP_011515506.1
XM_011517206.1 Intron XP_011515508.1
XM_017013716.1 Intron XP_016869205.1
Gene
ZFAT-AS1
Gene Name
ZFAT antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details