Product Details

SNP ID
rs2236582
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:109444615 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTCCTCTGGCTTCCTCCTGAATTGT[C/G]TCAGAGGTACACAGACTAGAGCATC
Phenotype
MIM: 607129 MIM: 603498
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MICAL1 PubMed Links

Gene Details

Gene
MICAL1
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159291.1 Intron NP_001152763.1
NM_001286613.1 Intron NP_001273542.1
NM_022765.3 Intron NP_073602.3
Gene
PPIL6
Gene Name
peptidylprolyl isomerase like 6
There are no transcripts associated with this gene.

Gene
SMPD2
Gene Name
sphingomyelin phosphodiesterase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003080.2 Intron NP_003071.2
XM_005267109.1 Intron XP_005267166.1
XM_011536079.1 Intron XP_011534381.1

View Full Product Details